Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RNF112 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007148.4 | NP_009079.2 | 14 | 2942 | ||
XM_006721571.3 | XP_006721634.1 | 14 | 6974 | |||
XM_006721572.3 | XP_006721635.1 | 14 | 6972 | |||
XM_006721573.3 | XP_006721636.1 | 14 | 2707 | |||
GenBank mRNA | AB026054.1 | - | 14 | 2799 | ||
AK226052.1 | - | 14 | 2882 | |||
BC053989.1 | - | 14 | 2857 | 5922474 |