Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SLMAP SIRNA Assays ›
Gene Symbol:
SLMAP
Gene Name:
sarcolemma associated protein
Gene Aliases:
SLAP
Chromosome Location:
Chr.3: 57756221 - 57930013 on Build GRCh38
UniGene ID:
Hs.476432
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139738
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SLMAP
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001304420.2
NP_001291349.1
18
2065
NM_001304421.2
NP_001291350.1
16
1951
NM_001304422.2
NP_001291351.1
6
547
NM_001304423.2
NP_001291352.1
4
349
NM_001311178.1
NP_001298107.1
6
547
NM_001311179.1
NP_001298108.1
5
424
NM_007159.4
NP_009090.2
17
2014
XM_005265456.3
XP_005265513.1
20
2952
XM_005265457.3
XP_005265514.1
20
2946
XM_005265458.3
XP_005265515.1
20
2952
XM_005265459.3
XP_005265516.1
19
2901
XM_005265460.3
XP_005265517.1
19
2901
XM_005265461.3
XP_005265518.1
19
2895
XM_005265462.3
XP_005265519.1
19
2889
XM_005265464.3
XP_005265521.1
19
2883
XM_005265465.3
XP_005265522.1
18
2850
XM_005265466.3
XP_005265523.1
18
2844
XM_005265467.3
XP_005265524.1
18
2838
XM_005265468.3
XP_005265525.1
19
2829
XM_005265470.3
XP_005265527.1
17
2787
XM_005265471.3
XP_005265528.1
17
2781
XM_005265472.3
XP_005265529.1
17
2715
XM_006713326.2
XP_006713389.1
17
2721
XM_011534103.1
XP_011532405.1
18
2838
XM_011534104.1
XP_011532406.1
17
2727
XM_011534105.1
XP_011532407.1
16
2664
XM_011534106.1
XP_011532408.1
6
422
XM_017007146.1
XP_016862635.1
20
2946
XM_017007147.1
XP_016862636.1
19
2895
XM_017007148.1
XP_016862637.1
19
2901
XM_017007149.1
XP_016862638.1
19
2895
XM_017007150.1
XP_016862639.1
19
2895
XM_017007151.1
XP_016862640.1
18
2850
XM_017007152.1
XP_016862641.1
18
2844
XM_017007153.1
XP_016862642.1
18
2832
XM_017007154.1
XP_016862643.1
18
2832
XM_017007155.1
XP_016862644.1
19
2823
XM_017007156.1
XP_016862645.1
19
2829
XM_017007157.1
XP_016862646.1
19
2823
XM_017007158.1
XP_016862647.1
17
2787
XM_017007159.1
XP_016862648.1
18
2778
XM_017007160.1
XP_016862649.1
17
2781
XM_017007161.1
XP_016862650.1
18
2772
XM_017007162.1
XP_016862651.1
18
2778
XM_017007163.1
XP_016862652.1
18
2772
XM_017007164.1
XP_016862653.1
17
2721
XM_017007165.1
XP_016862654.1
17
2709
XM_017007166.1
XP_016862655.1
17
2709
XM_017007167.1
XP_016862656.1
16
2664
XM_017007168.1
XP_016862657.1
16
2658
XM_017007170.1
XP_016862659.1
6
422
GenBank mRNA
AB046821.1
-
15
1255
AF100750.1
-
6
491
AF304450.1
-
6
420
AK300106.1
-
4
272
AK302934.1
-
5
299
AK304493.1
-
6
474
AK316436.1
-
6
415
AL834538.1
-
2
88
BC114627.1
-
16
1649
40081892
BC115701.1
-
18
1763
40081893
CR627321.1
-
5
281
DB100357.1
-
6
551
Back To Top
More Information
Panther Classification:
Molecular Function -
scaffold/adaptor protein
Gene Ontology Categories:
Function(s)
Process(es)
muscle contraction
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-97l6p:80/100.66.76.85:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline