Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHOC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001269039.2 | NP_001255968.1 | 2 | 780 | ||
NM_001324336.1 | NP_001311265.1 | 2 | 737 | |||
NM_001324337.1 | NP_001311266.1 | 3 | 965 | |||
NM_007373.3 | NP_031399.2 | 2 | 780 | |||
XM_017016702.1 | XP_016872191.1 | 2 | 685 | |||
XM_017016703.1 | XP_016872192.1 | 2 | 6502 | |||
XM_017016704.1 | XP_016872193.1 | 2 | 684 | |||
GenBank mRNA | AB020669.1 | - | 2 | 708 | ||
AF054828.1 | - | 2 | 702 | |||
AF068920.1 | - | 1 | 431 | |||
AK292833.1 | - | 3 | 906 | |||
BC044752.1 | - | 2 | 716 | |||
BC050445.1 | - | 1 | 635 | 6067669 | ||
BX649055.1 | - | 2, 3 | 548 |