Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282902.2 | NP_001269831.1 | 2 | 461 | ||
NM_001282903.2 | NP_001269832.1 | 3 | 950 | |||
NM_014112.4 | NP_054831.2 | 3 | 964 | |||
XM_005251049.2 | XP_005251106.1 | 2 | 779 | |||
XM_006716625.1 | XP_006716688.1 | 3 | 901 | |||
XM_011517264.1 | XP_011515566.1 | 4 | 1034 | |||
XM_011517266.2 | XP_011515568.1 | 3 | 769 | |||
XM_011517267.1 | XP_011515569.1 | 3 | 887 | |||
XM_011517268.1 | XP_011515570.1 | 3 | 876 | |||
XM_017013801.1 | XP_016869290.1 | 2 | 4563 | |||
GenBank mRNA | AF183810.1 | - | 3 | 958 | ||
AF264784.1 | - | 2 | 505 | |||
AK304046.1 | - | 2 | 456 | |||
AK309357.1 | - | 2 | 675 | |||
BC125020.1 | - | 3 | 794 | 40073132 | ||
DC339831.1 | - | 2 | 461 |