Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AAAS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001173466.1 | NP_001166937.1 | 15 | 1600 | ||
NM_015665.5 | NP_056480.1 | 16 | 1699 | |||
XM_011538778.1 | XP_011537080.1 | 16 | 1673 | |||
XM_011538780.1 | XP_011537082.1 | 15 | 1574 | |||
GenBank mRNA | AF226048.1 | - | 16 | 1618 | ||
AJ289857.1 | - | 16 | 1626 | |||
AK000833.1 | - | 16 | 1626 | |||
AK094302.1 | - | 14 | 1879 | |||
AK222509.1 | - | 16 | 1626 | |||
AK293342.1 | - | 15 | 1557 | |||
AL110160.1 | - | 10 | 909 | |||
AY237818.1 | - | 15 | 1568 | |||
BC000659.2 | - | 16 | 1661 | 3347423 | ||
BC018836.2 | - | 16 | 1661 | |||
BT006912.1 | - | 16 | 1536 |