Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003864.3 | NP_003855.1 | 3 | 763 | ||
GenBank mRNA | AF055993.1 | - | 3 | 762 | ||
BC016757.1 | - | 3 | 758 | 4074154 |