Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256279.1 | NP_001243208.1 | 5 | 2170 | ||
NM_001256280.1 | NP_001243209.1 | 3 | 1975 | |||
NM_019591.3 | NP_062537.2 | 4 | 2071 | |||
XM_005266182.3 | XP_005266239.1 | 5 | 2158 | |||
XM_005266183.3 | XP_005266240.1 | 6 | 2361 | |||
XM_011534829.1 | XP_011533131.1 | 5 | 2274 | |||
XM_017019920.1 | XP_016875409.1 | 6 | 2794 | |||
XM_017019921.1 | XP_016875410.1 | 6 | 2257 | |||
XM_017019922.1 | XP_016875411.1 | 5 | 3143 | |||
XM_017019923.1 | XP_016875412.1 | 5 | 3074 | |||
GenBank mRNA | AK000762.1 | - | 1 | 1274 | ||
AK122941.1 | - | 3 | 4184 | |||
AK225296.1 | - | 1 | 1274 | |||
AK291243.1 | - | 4 | 2038 | |||
AL833057.1 | - | 1 | 848 | |||
BC008211.1 | - | 1 | 811 | |||
BC046206.2 | - | 4 | 1972 | 6176427 | ||
BT006963.1 | - | 1 | 576 | |||
EU668356.1 | - | 4 | 1949 |