Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020638.2 | NP_065689.1 | 3 | 2251 | ||
GenBank mRNA | AB047858.1 | - | 3 | 2237 |