Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TNFAIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021137.4 | NP_066960.1 | 7 | 2788 | ||
XM_017024993.1 | XP_016880482.1 | 7 | 2866 | |||
GenBank mRNA | BC003694.2 | - | 7 | 2552 | ||
M80783.1 | - | 7 | 2494 |