Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184879.1 | NP_001171808.1 | 8 | 2940 | ||
NM_001184881.1 | NP_001171810.1 | 6 | 2791 | |||
NM_001184882.1 | NP_001171811.1 | 6 | 2547 | |||
NM_003874.3 | NP_003865.1 | 7 | 2889 | |||
XM_011510094.2 | XP_011508396.1 | 8 | 2903 | |||
XM_011510095.2 | XP_011508397.1 | 7 | 2579 | |||
GenBank mRNA | AJ223324.1 | - | 7 | 2854 | ||
BC020063.1 | - | 7 | 2834 | 4385965 | ||
CR933666.1 | - | 6 | 2508 |