Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021160.2 | NP_066983.1 | 1, 2 | 212 | ||
GenBank mRNA | AF193047.2 | - | 1, 2 | 161 | ||
AK023194.1 | - | 1, 2 | 163 | |||
AK289469.1 | - | 1, 2 | 208 | |||
AK303496.1 | - | 1, 2 | 163 | |||
BC031839.1 | - | 1, 2 | 167 | 4931559 | ||
DC360732.1 | - | 1, 2 | 212 |