Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABHD16A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177515.1 | NP_001170986.1 | 3, 4 | 645 | ||
NM_021160.2 | NP_066983.1 | 5, 6 | 518 | |||
NR_033488.1 | 5, 6 | 769 | ||||
NR_033489.1 | 3, 4 | 394 | ||||
GenBank mRNA | AF193047.2 | - | 5, 6 | 467 | ||
AK023194.1 | - | 5, 6 | 469 | |||
AK055255.1 | - | 5, 6 | 769 | |||
AK289469.1 | - | 5, 6 | 514 | |||
AK297712.1 | - | 3, 4 | 642 | |||
AK303496.1 | - | 4, 5 | 402 | |||
BC031839.1 | - | 5, 6 | 473 | 4931559 | ||
BP261875.1 | - | 3, 4 | 438 | |||
DC318775.1 | - | 3, 4 | 390 | |||
DC360732.1 | - | 5, 6 | 518 |