Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018412.3 | NP_060882.2 | 5 | 691 | ||
NM_021908.2 | NP_068708.1 | 5 | 691 | |||
GenBank mRNA | AF234882.1 | - | 5 | 690 | ||
AF234883.1 | - | 5 | 690 | |||
AF415174.1 | - | 3 | 118 | |||
AJ277291.1 | - | 4 | 326 | |||
AK093212.1 | - | 6 | 940 | |||
AK292372.1 | - | 5 | 517 | |||
AK297510.1 | - | 5 | 568 | |||
AK297861.1 | - | 5 | 399 | |||
AK298540.1 | - | 5 | 513 | |||
AY009152.1 | - | 5 | 682 | |||
AY009153.1 | - | 5 | 682 | |||
BC030954.1 | - | 6 | 496 | |||
BC075855.1 | - | 5 | 691 | 5562032 | ||
KJ534947.1 | - | 5 | 477 |