Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021998.4 | NP_068838.3 | 9 | 2263 | ||
XM_005262186.2 | XP_005262243.1 | 11 | 2826 | |||
XM_005262187.1 | XP_005262244.1 | 11 | 2715 | |||
XM_005262188.1 | XP_005262245.1 | 10 | 2401 | |||
XM_005262189.2 | XP_005262246.1 | 10 | 2688 | |||
XM_011531019.1 | XP_011529321.1 | 10 | 3201 | |||
XM_011531020.1 | XP_011529322.1 | 11 | 2838 | |||
XM_011531021.1 | XP_011529323.1 | 11 | 2843 | |||
XM_011531022.1 | XP_011529324.1 | 11 | 2727 | |||
XM_011531023.2 | XP_011529325.1 | 10 | 2291 | |||
XM_011531024.1 | XP_011529326.1 | 10 | 3189 | |||
XM_011531025.1 | XP_011529327.1 | 9 | 3063 | |||
XM_011531026.1 | XP_011529328.1 | 9 | 3051 | |||
XM_017029803.1 | XP_016885292.1 | 8 | 3151 | |||
XM_017029804.1 | XP_016885293.1 | 11 | 2831 | |||
XM_017029805.1 | XP_016885294.1 | 9 | 2281 | |||
XM_017029806.1 | XP_016885295.1 | 7 | 3001 | |||
XM_017029807.1 | XP_016885296.1 | 10 | 2693 | |||
XM_017029808.1 | XP_016885297.1 | 10 | 2577 | |||
XM_017029809.1 | XP_016885298.1 | 8 | 2143 | |||
GenBank mRNA | AK294953.1 | - | 3 | 1643 | ||
AK299604.1 | - | 5 | 1174 | |||
AK299933.1 | - | 7 | 1503 | |||
BC067294.1 | - | 7 | 1947 | 30386278 | ||
BX648117.1 | - | 10 | 2940 |