Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TIA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022037.2 | NP_071320.2 | 12 | 1276 | ||
NM_022173.2 | NP_071505.2 | 13 | 1309 | |||
XM_005264527.1 | XP_005264584.1 | 12 | 1283 | |||
XM_005264528.4 | XP_005264585.1 | 12 | 1221 | |||
XM_005264531.1 | XP_005264588.1 | 14 | 1356 | |||
XM_011533077.1 | XP_011531379.1 | 13 | 1316 | |||
XM_011533078.1 | XP_011531380.1 | 12 | 4982 | |||
XM_011533081.2 | XP_011531383.1 | 13 | 2082 | |||
XM_011533082.2 | XP_011531384.1 | 11 | 5654 | |||
XM_017004789.1 | XP_016860278.1 | 13 | 1138 | |||
XM_017004790.1 | XP_016860279.1 | 12 | 1104 | |||
XM_017004791.1 | XP_016860280.1 | 11 | 1188 | |||
XM_017004792.1 | XP_016860281.1 | 12 | 3493 | |||
XM_017004793.1 | XP_016860282.1 | 12 | 3318 | |||
XM_017004794.1 | XP_016860283.1 | 12 | 2050 | |||
XM_017004795.1 | XP_016860284.1 | 14 | 1316 | |||
XM_017004796.1 | XP_016860285.1 | 12 | 4824 | |||
XM_017004797.1 | XP_016860286.1 | 13 | 2620 | |||
XM_017004798.1 | XP_016860287.1 | 13 | 1351 | |||
XM_017004799.1 | XP_016860288.1 | 13 | 1313 | |||
XM_017004800.1 | XP_016860289.1 | 12 | 1280 | |||
GenBank mRNA | AB209211.1 | - | 13 | 1450 | ||
DR157578.1 | - | 7 | 648 | |||
M77142.1 | - | 12 | 1115 |