Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FZD8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031866.2 | NP_114072.1 | 1 | 2636 | ||
GenBank mRNA | AB043703.1 | - | 1 | 2636 | ||
AF086500.1 | - | 1 | 481 |
Set Membership: |
G-protein coupled receptor |