Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004178.4 | NP_004169.3 | 8 | 822 | ||
NM_134323.1 | NP_599150.1 | 8 | 1287 | |||
NM_134324.2 | NP_599151.2 | 8 | 862 | |||
XM_005269114.1 | XP_005269171.1 | 8 | 857 | |||
XM_005269115.2 | XP_005269172.1 | 8 | 864 | |||
XM_005269117.1 | XP_005269174.1 | 7 | 778 | |||
XM_005269120.4 | XP_005269177.1 | 8 | 1270 | |||
XM_005269122.3 | XP_005269179.1 | 8 | 842 | |||
XM_006719581.1 | XP_006719644.1 | 7 | 706 | |||
XM_011538712.2 | XP_011537014.1 | 8 | 911 | |||
XM_017019910.1 | XP_016875399.1 | 7 | 729 | |||
XM_017019911.1 | XP_016875400.1 | 8 | 1000 | |||
GenBank mRNA | AK290737.1 | - | 8 | 860 | ||
BC002419.2 | - | 8 | 1270 | |||
BC005860.2 | - | 8 | 1152 | 2960701 | ||
BT007140.1 | - | 8 | 799 | |||
M60801.1 | - | 8 | 790 | |||
U08998.2 | - | 8 | 887 |