Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TARS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258437.1 | NP_001245366.1 | 3, 4 | 278 | ||
NM_001258438.1 | NP_001245367.1 | 2, 3 | 452 | |||
NM_152295.4 | NP_689508.3 | 2, 3 | 452 | |||
NR_047677.1 | 2, 3 | 452 | ||||
GenBank mRNA | AK222804.1 | - | 2, 3 | 209 | ||
AK292346.1 | - | 3, 4 | 278 | |||
AK293620.1 | - | 2, 3 | 263 | |||
AK296783.1 | - | 2, 3 | 251 | |||
AK311361.1 | - | 263 | ||||
AK315713.1 | - | 2, 3 | 263 | |||
BC000517.2 | - | 2, 3 | 221 | 2822851 | ||
BC010578.1 | - | 2, 3 | 239 | 3458845 | ||
BC088355.1 | - | 2, 3 | 230 | |||
M63180.1 | - | 2, 3 | 243 |