Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGKD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003648.2 | NP_003639.2 | 1, 2 | 215 | ||
NM_152879.2 | NP_690618.2 | 2, 3 | 280 | |||
XM_011512028.2 | XP_011510330.1 | 2, 3 | 1739 | |||
XM_011512029.2 | XP_011510331.1 | 2, 3 | 1740 | |||
XM_011512030.2 | XP_011510332.1 | 2, 3 | 336 | |||
XM_011512031.2 | XP_011510333.1 | 2, 3 | 210 | |||
XM_011512032.2 | XP_011510334.1 | 2, 3 | 194 | |||
XM_011512033.2 | XP_011510335.1 | 2, 3 | 1741 | |||
XM_011512039.2 | XP_011510341.1 | 2, 3 | 1741 | |||
XM_017005138.1 | XP_016860627.1 | 2, 3 | 618 | |||
GenBank mRNA | AB078966.1 | - | 2, 3 | 280 | ||
D63479.3 | - | 2, 3 | 221 | |||
D73409.1 | - | 1, 2 | 216 |