Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141919.1 | NP_001135391.1 | 4 | 372 | ||
NM_001141920.1 | NP_001135392.1 | 4 | 372 | |||
NM_175569.2 | NP_780778.1 | 4 | 372 | |||
XM_005274587.3 | XP_005274644.1 | 4 | 1300 | |||
XM_011545575.2 | XP_011543877.1 | 4 | 1316 | |||
XM_017029787.1 | XP_016885276.1 | 3 | 1272 | |||
GenBank mRNA | AF380356.1 | - | 4 | 350 | ||
AF534880.1 | - | 4 | 346 | |||
AK304164.1 | - | 4 | 372 | |||
BC100764.2 | - | 5 | 337 | |||
BC100765.2 | - | 4 | 234 | 40000158 | ||
BC100766.2 | - | 4 | 235 | 40000159 | ||
BC100767.1 | - | 4 | 234 | 40000160 |