Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF227 SIRNA Assays ›
Gene Symbol:
ZNF227
Gene Name:
zinc finger protein 227
Gene Aliases:
-
Chromosome Location:
Chr.19: 44207527 - 44237268 on Build GRCh38
UniGene ID:
Hs.298089
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
139990
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF227
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001289166.1
NP_001276095.1
6
647
NM_001289167.1
NP_001276096.1
6
603
NM_001289168.1
NP_001276097.1
5
582
NM_001289169.1
NP_001276098.1
5
585
NM_001289170.1
NP_001276099.1
4
498
NM_001289171.1
NP_001276100.1
4
501
NM_001289172.1
NP_001276101.1
4
559
NM_001289173.1
NP_001276102.1
5
581
NM_182490.2
NP_872296.1
6
644
XM_005259232.2
XP_005259289.1
6
775
XM_006723371.1
XP_006723434.1
5
556
XM_011527289.2
XP_011525591.1
6
923
XM_011527290.1
XP_011525592.1
6
619
XM_017027266.1
XP_016882755.1
6
778
XM_017027267.1
XP_016882756.1
5
659
XM_017027268.1
XP_016882757.1
5
713
XM_017027269.1
XP_016882758.1
5
683
XM_017027270.1
XP_016882759.1
5
861
XM_017027271.1
XP_016882760.1
5
859
XM_017027272.1
XP_016882761.1
5
524
XM_017027273.1
XP_016882762.1
5
598
XM_017027274.1
XP_016882763.1
5
863
XM_017027275.1
XP_016882764.1
6
746
XM_017027276.1
XP_016882765.1
6
742
XM_017027277.1
XP_016882766.1
5
606
XM_017027278.1
XP_016882767.1
3
240
XM_017027279.1
XP_016882768.1
4
632
GenBank mRNA
AK092253.1
-
6
644
AK299266.1
-
4
535
AK300576.1
-
4
456
AK316048.1
-
6
593
AL833012.1
-
4
402
AY166786.1
-
3
294
BC047570.1
-
6
634
4838846
BU153285.1
-
4
537
DC341721.1
-
4
461
Back To Top
More Information
Panther Classification:
Molecular Function -
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0