Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF227 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289166.1 | NP_001276095.1 | 6 | 2909 | ||
NM_001289167.1 | NP_001276096.1 | 6 | 2865 | |||
NM_001289168.1 | NP_001276097.1 | 5 | 2844 | |||
NM_001289169.1 | NP_001276098.1 | 5 | 2847 | |||
NM_001289170.1 | NP_001276099.1 | 4 | 2760 | |||
NM_001289171.1 | NP_001276100.1 | 4 | 2763 | |||
NM_001289172.1 | NP_001276101.1 | 4 | 2821 | |||
NM_001289173.1 | NP_001276102.1 | 5 | 2843 | |||
NM_182490.2 | NP_872296.1 | 6 | 2906 | |||
XM_006723371.1 | XP_006723434.1 | 5 | 2818 | |||
XM_011527290.1 | XP_011525592.1 | 6 | 2881 | |||
XM_017027267.1 | XP_016882756.1 | 5 | 2921 | |||
XM_017027272.1 | XP_016882761.1 | 5 | 2786 | |||
XM_017027273.1 | XP_016882762.1 | 5 | 2860 | |||
XM_017027274.1 | XP_016882763.1 | 5 | 3125 | |||
XM_017027279.1 | XP_016882768.1 | 4 | 2894 | |||
GenBank mRNA | AK092253.1 | - | 6 | 2906 | ||
AK299266.1 | - | 4 | 2797 | |||
AL833012.1 | - | 4 | 2664 | |||
AY500355.1 | - | 2 | 1611 | |||
BC047570.1 | - | 6 | 2896 | 4838846 |