Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243891.1 | NP_001230820.1 | 7 | 1315 | ||
NM_001243892.1 | NP_001230821.1 | 6 | 1261 | |||
NM_001243894.1 | NP_001230823.1 | 5 | 1228 | |||
NM_001243895.1 | NP_001230824.1 | 6 | 1341 | |||
NM_003288.3 | NP_003279.2 | 7 | 1390 | |||
NM_199359.2 | NP_955391.1 | 6 | 1330 | |||
NM_199360.2 | NP_955392.1 | 9 | 1459 | |||
NM_199361.2 | NP_955393.1 | 8 | 1399 | |||
NM_199362.2 | NP_955394.1 | 8 | 1432 | |||
NM_199363.2 | NP_955395.1 | 7 | 1372 | |||
NR_045090.1 | 7 | 1359 | ||||
GenBank mRNA | AB062396.1 | - | 3 | 848 | ||
AF004429.1 | - | 4 | 837 | |||
AF004430.1 | - | 7 | 1328 | |||
AK055068.1 | - | 7 | 1331 | |||
AK125709.1 | - | 1 | 1285 | |||
AK128744.1 | - | 2 | 3085 | |||
AK223031.1 | - | 7 | 1302 | |||
AK293349.1 | - | 7 | 1212 | |||
AK297064.1 | - | 6 | 1288 | |||
BC006804.1 | - | 7 | 1275 | 3446037 | ||
CR749208.1 | - | 6 | 1229 |