Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPD52L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243891.1 | NP_001230820.1 | 6, 7 | 589 | ||
NM_001243892.1 | NP_001230821.1 | 5, 6 | 535 | |||
NM_001243894.1 | NP_001230823.1 | 4, 5 | 502 | |||
NM_003288.3 | NP_003279.2 | 6, 7 | 664 | |||
NM_199359.2 | NP_955391.1 | 5, 6 | 604 | |||
NM_199360.2 | NP_955392.1 | 8, 9 | 733 | |||
NM_199361.2 | NP_955393.1 | 7, 8 | 673 | |||
NM_199362.2 | NP_955394.1 | 7, 8 | 706 | |||
NM_199363.2 | NP_955395.1 | 6, 7 | 646 | |||
NR_045090.1 | 6, 7 | 633 | ||||
GenBank mRNA | AB062396.1 | - | 2, 3 | 122 | ||
AF004429.1 | - | 3, 4 | 111 | |||
AF004430.1 | - | 6, 7 | 602 | |||
AK055068.1 | - | 6, 7 | 605 | |||
AK128744.1 | - | 1, 2 | 2358 | |||
AK223031.1 | - | 6, 7 | 576 | |||
AK293349.1 | - | 6, 7 | 486 | |||
AK297064.1 | - | 5, 6 | 562 | |||
AK298366.1 | - | 5, 6 | 520 | |||
AK314501.1 | - | 6, 7 | 622 | |||
AU124850.1 | - | 6, 7 | 593 | |||
AY323819.1 | - | 6, 7 | 533 | |||
BC006804.1 | - | 6, 7 | 549 | 3446037 | ||
BI667351.1 | - | 7, 8 | 485 | |||
BI754201.1 | - | 7, 8 | 560 | |||
BM473873.1 | - | 6, 7 | 621 | |||
BQ948007.1 | - | 6, 7 | 559 | |||
BT019631.1 | - | 7, 8 | 568 | |||
CR542036.1 | - | 6, 7 | 526 | |||
CR749208.1 | - | 5, 6 | 503 | |||
DA689198.1 | - | 4, 5 | 401 | |||
DC362382.1 | - | 5, 6 | 520 | |||
DC428780.1 | - | 6, 7 | 519 |