Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH2B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005475.2 | NP_005466.1 | 2 | 445 | ||
XM_005253818.4 | XP_005253875.1 | 2 | 469 | |||
XM_005253819.4 | XP_005253876.1 | 2 | 469 | |||
XM_011537719.2 | XP_011536021.1 | 2 | 469 | |||
XM_011537720.2 | XP_011536022.1 | 2 | 134 | |||
GenBank mRNA | AF055581.1 | - | 2 | 445 | ||
AJ012793.1 | - | 1 | 88 | |||
BC136451.1 | - | 2 | 131 | 9020440 |