Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144823.2 | NP_001138295.1 | 3 | 514 | ||
NM_001320835.1 | NP_001307764.1 | 3 | 514 | |||
NM_005848.4 | NP_005839.3 | 3 | 507 | |||
XM_005254120.4 | XP_005254177.1 | 2 | 293 | |||
XM_005254121.3 | XP_005254178.1 | 3 | 514 | |||
XM_005254122.4 | XP_005254179.1 | 3 | 373 | |||
XM_006720371.3 | XP_006720434.1 | 3 | 249 | |||
XM_011521156.2 | XP_011519458.1 | 3 | 509 | |||
XM_017021862.1 | XP_016877351.1 | 2 | 234 | |||
XM_017021863.1 | XP_016877352.1 | 2 | 292 | |||
XM_017021864.1 | XP_016877353.1 | 3 | 509 | |||
XM_017021865.1 | XP_016877354.1 | 3 | 372 | |||
GenBank mRNA | AF534403.1 | - | 3 | 469 | ||
AK091368.1 | - | 3 | 469 | |||
AK290292.1 | - | 3 | 322 | |||
AK307411.1 | - | 3 | 514 | |||
AL832602.2 | - | 1 | 33 | |||
AL833317.1 | - | 3 | 345 | |||
BC028057.1 | - | 3 | 359 | |||
BC041706.1 | - | 3 | 338 | 5530033 |