Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144823.2 | NP_001138295.1 | 3 | 616 | ||
NM_001320835.1 | NP_001307764.1 | 3 | 616 | |||
NM_005848.4 | NP_005839.3 | 3 | 609 | |||
XM_005254120.4 | XP_005254177.1 | 2 | 395 | |||
XM_005254121.3 | XP_005254178.1 | 3 | 616 | |||
XM_005254122.4 | XP_005254179.1 | 3 | 475 | |||
XM_006720371.3 | XP_006720434.1 | 3 | 351 | |||
XM_011521156.2 | XP_011519458.1 | 3 | 611 | |||
XM_017021862.1 | XP_016877351.1 | 2 | 336 | |||
XM_017021863.1 | XP_016877352.1 | 2 | 394 | |||
XM_017021864.1 | XP_016877353.1 | 3 | 611 | |||
XM_017021865.1 | XP_016877354.1 | 3 | 474 | |||
GenBank mRNA | AF534403.1 | - | 3 | 571 | ||
AK091368.1 | - | 3 | 571 | |||
AK290292.1 | - | 3 | 424 | |||
AK299570.1 | - | 3 | 530 | |||
AK307411.1 | - | 3 | 616 | |||
AL832602.2 | - | 1 | 135 | |||
AL833317.1 | - | 3 | 447 | |||
BC028057.1 | - | 3 | 461 | |||
BC041706.1 | - | 3 | 440 | 5530033 |