Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321445.1 | NP_001308374.1 | 4 | 579 | ||
NM_006815.3 | NP_006806.1 | 3 | 560 | |||
GenBank mRNA | AK298167.1 | - | 3 | 454 | ||
AK303189.1 | - | 4 | 384 | |||
AK312975.1 | - | 3 | 535 | |||
BC025957.1 | - | 3 | 475 | 5020405 | ||
BI086507.1 | - | 3 | 488 | |||
CR541663.1 | - | 3 | 452 | |||
CR541682.1 | - | 3 | 452 | |||
X92098.1 | - | 3 | 479 |