Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMED10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006827.5 | NP_006818.3 | 3 | 432 | ||
GenBank mRNA | AJ004913.1 | - | 3 | 376 | ||
AK222972.1 | - | 3 | 384 | |||
AK296800.1 | - | 3 | 401 | |||
AK302920.1 | - | 2 | 93 | |||
AK308864.1 | - | 1 | 51 | |||
AK310371.1 | - | 3 | 383 | |||
AK312384.1 | - | 3 | 401 | |||
AL832012.2 | - | 3 | 376 | |||
BC001496.2 | - | 3 | 417 | 2959718 | ||
BC001825.2 | - | 3 | 417 | 2959718 | ||
BX248754.1 | - | 3 | 383 | |||
BX504393.1 | - | 3 | 433 | |||
X97442.1 | - | 3 | 376 |