Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNF8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317192.1 | NP_001304121.1 | 8 | 1103 | ||
NM_001317193.1 | NP_001304122.1 | 7 | 1055 | |||
NM_001317194.1 | NP_001304123.1 | 10 | 1196 | |||
NM_007241.3 | NP_009172.2 | 8 | 1106 | |||
NR_133679.1 | 8 | 1145 | ||||
GenBank mRNA | BC008976.1 | - | 8 | 1056 | 3452153 |