Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFT81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143779.1 | NP_001137251.1 | 2 | 212 | ||
NM_014055.3 | NP_054774.2 | 2 | 630 | |||
NM_031473.3 | NP_113661.2 | 2 | 630 | |||
XM_011538232.2 | XP_011536534.1 | 2 | 158 | |||
XM_017019217.1 | XP_016874706.1 | 2 | 322 | |||
XM_017019218.1 | XP_016874707.1 | 2 | 630 | |||
GenBank mRNA | AF139540.1 | - | 2 | 170 | ||
AF250326.1 | - | 1 | 123 | |||
AF332010.1 | - | 2 | 588 | |||
AK291043.1 | - | 2 | 532 | |||
BC004536.2 | - | 2 | 466 | 3608705 | ||
BC005396.1 | - | 2 | 132 | |||
BC108257.1 | - | 1 | 95 | 6156440 | ||
BP351086.1 | - | 2 | 212 |