Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC113 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142302.1 | NP_001135774.1 | 8 | 1615 | ||
NM_014157.3 | NP_054876.2 | 9 | 1777 | |||
XM_011523038.2 | XP_011521340.1 | 9 | 1632 | |||
XM_011523039.2 | XP_011521341.1 | 11 | 2525 | |||
XM_011523040.2 | XP_011521342.1 | 9 | 2050 | |||
XM_011523041.2 | XP_011521343.1 | 9 | 1618 | |||
XM_017023176.1 | XP_016878665.1 | 10 | 1940 | |||
XM_017023177.1 | XP_016878666.1 | 8 | 1634 | |||
GenBank mRNA | AL136785.1 | - | 9 | 1754 | ||
BC126445.1 | - | 9 | 1746 | 8992161 | ||
BC126447.1 | - | 9 | 1746 | 8992163 |