Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014167.4 | NP_054886.2 | 2 | 374 | ||
NR_033192.1 | 2 | 303 | ||||
GenBank mRNA | AF161477.1 | - | 2 | 348 | ||
AF213377.1 | - | 2 | 341 | |||
AK001156.1 | - | 2 | 350 | |||
AK225139.1 | - | 2 | 353 | |||
BC020647.1 | - | 2 | 337 | 4771954 | ||
BG538660.1 | - | 2 | 379 | |||
BM846580.1 | - | 2 | 273 | |||
DA666239.1 | - | 2 | 303 |