Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014167.4 | NP_054886.2 | 4 | 1023 | ||
NR_033192.1 | 4 | 952 | ||||
GenBank mRNA | AF161477.1 | - | 4 | 997 | ||
AF213377.1 | - | 4 | 990 | |||
AL832749.1 | - | 2 | 2043 | |||
BC020647.1 | - | 4 | 986 | 4771954 |