Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MBD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281453.1 | NP_001268382.1 | 7 | 1425 | ||
NM_001281454.1 | NP_001268383.1 | 7 | 1329 | |||
GenBank mRNA | AF072247.1 | - | 7 | 1318 | ||
AK001474.1 | - | 6 | 1125 | |||
AK021532.1 | - | 6 | 1076 | |||
BC009372.1 | - | 7 | 1261 | 4122128 | ||
BC009438.1 | - | 4 | 1120 | |||
BC018784.1 | - | 1 | 428 | |||
BC032443.1 | - | 5 | 933 | |||
BC043619.1 | - | 7 | 1279 | 6156267 |