Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EDC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014329.4 | NP_055144.3 | 13 | 1768 | ||
GenBank mRNA | AK128582.1 | - | 12 | 2242 | ||
AK291049.1 | - | 13 | 1536 | |||
AK302563.1 | - | 13 | 1534 | |||
BC043616.1 | - | 13 | 1671 | 6065418 | ||
BC053598.1 | - | 8 | 908 | |||
BC064567.1 | - | 13 | 1665 | 5575195 | ||
L26339.1 | - | 13 | 1665 | |||
U17474.1 | - | 3 | 305 |