Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278241.1 | NP_001265170.1 | 12 | 1545 | ||
NM_014389.2 | NP_055204.3 | 11 | 1463 | |||
GenBank mRNA | AF547989.1 | - | 11 | 1237 | ||
AK226056.1 | - | 11 | 1243 | |||
AK291163.1 | - | 11 | 1263 | |||
AK293838.1 | - | 12 | 1332 | |||
AK299084.1 | - | 11 | 1451 | |||
AY882602.1 | - | 11 | 1030 | |||
BC002875.2 | - | 1 | 78 | |||
BC010457.2 | - | 10 | 991 | |||
BC069058.1 | - | 11 | 1237 | 6068755 | ||
U88153.2 | - | 10 | 1672 | |||
U88154.1 | - | 11 | 1144 |