Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PELP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278241.1 | NP_001265170.1 | 13 | 1606 | ||
NM_014389.2 | NP_055204.3 | 12 | 1524 | |||
GenBank mRNA | AF547989.1 | - | 12 | 1298 | ||
AK226056.1 | - | 12 | 1304 | |||
AK291163.1 | - | 12 | 1324 | |||
AK293838.1 | - | 13 | 1393 | |||
AK299084.1 | - | 12 | 1512 | |||
AY882602.1 | - | 12 | 1091 | |||
BC002875.2 | - | 2 | 139 | |||
BC010457.2 | - | 11 | 1052 | |||
BC069058.1 | - | 12 | 1298 | 6068755 | ||
U88153.2 | - | 11 | 1733 | |||
U88154.1 | - | 12 | 1205 |