Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDR2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014603.2 | NP_055418.2 | 5 | 1790 | ||
XM_006721852.2 | XP_006721915.1 | 5 | 1748 | |||
GenBank mRNA | AK294006.1 | - | 5 | 1790 | ||
BC013105.2 | - | 1 | 332 | |||
BC047534.1 | - | 5 | 1790 | 5288296 | ||
L02867.1 | - | 5 | 1588 |