Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDR2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014603.2 | NP_055418.2 | 5 | 3457 | ||
XM_006721852.2 | XP_006721915.1 | 5 | 3415 | |||
GenBank mRNA | BC013105.2 | - | 1 | 1996 | ||
BC047534.1 | - | 5 | 3456 | 5288296 | ||
CR749715.1 | - | 1 | 766 |