Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001303468.1 | NP_001290397.1 | 9 | 914 | ||
NM_003930.4 | NP_003921.2 | 9 | 1022 | |||
XM_017012771.1 | XP_016868260.1 | 9 | 984 | |||
GenBank mRNA | AB014486.1 | - | 9 | 741 | ||
AF051323.1 | - | 9 | 880 | |||
AF072166.1 | - | 9 | 708 | |||
AJ004886.1 | - | 9 | 708 | |||
AK222885.1 | - | 9 | 868 | |||
AK299216.1 | - | 10 | 861 | |||
AK299317.1 | - | 9 | 811 | |||
AK313209.1 | - | 9 | 868 | |||
BC002893.2 | - | 9 | 866 | 3940826 | ||
BC036044.1 | - | 9 | 869 | 5272981 |