Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB11FIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142272.1 | NP_001135744.1 | 12 | 2851 | ||
NM_014700.3 | NP_055515.1 | 14 | 3897 | |||
XM_005255713.3 | XP_005255770.1 | 15 | 4067 | |||
XM_005255714.1 | XP_005255771.1 | 13 | 2912 | |||
XM_005255715.4 | XP_005255772.1 | 13 | 3718 | |||
XM_011522764.2 | XP_011521066.1 | 13 | 2944 | |||
XM_011522765.2 | XP_011521067.1 | 13 | 2770 | |||
GenBank mRNA | AB014565.1 | - | 14 | 3945 | ||
AF038180.1 | - | 1 | 1368 | |||
AF395731.1 | - | 14 | 3945 | |||
AY130007.1 | - | 6 | 1992 | |||
BC051360.1 | - | 14 | 3896 | 6646253 |