Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCP110 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199022.2 | NP_001185951.2 | 3 | 484 | ||
NM_001323569.1 | NP_001310498.1 | 4 | 627 | |||
NM_001323570.1 | NP_001310499.1 | 5 | 1072 | |||
NM_001323571.1 | NP_001310500.1 | 5 | 867 | |||
NM_001323572.1 | NP_001310501.1 | 3 | 484 | |||
NM_001323576.1 | NP_001310505.1 | 4 | 627 | |||
NM_001323577.1 | NP_001310506.1 | 3 | 484 | |||
NM_014711.5 | NP_055526.4 | 4 | 627 | |||
XM_011545991.2 | XP_011544293.1 | 5 | 1050 | |||
XM_011545992.2 | XP_011544294.1 | 5 | 768 | |||
XM_017023908.1 | XP_016879397.1 | 4 | 1544 | |||
GenBank mRNA | AB007879.1 | - | 4 | 528 | ||
AK300426.1 | - | 3 | 397 | |||
BC036654.2 | - | 4 | 598 | 5267904 | ||
BY798911.2 | - | 3 | 484 | |||
CR749255.1 | - | 3 | 384 |