Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IST1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270975.1 | NP_001257904.1 | 10 | 1235 | ||
NM_001270976.1 | NP_001257905.1 | 11 | 1350 | |||
NM_001270977.1 | NP_001257906.1 | 9 | 1142 | |||
NM_001270978.1 | NP_001257907.1 | 7 | 863 | |||
NM_001270979.1 | NP_001257908.1 | 6 | 779 | |||
NM_014761.3 | NP_055576.2 | 10 | 1231 | |||
GenBank mRNA | AB097052.1 | - | 10 | 1180 | ||
AK057902.1 | - | 10 | 1198 | |||
AK291388.1 | - | 10 | 1214 | |||
AK293022.1 | - | 10 | 1196 | |||
AK293040.1 | - | 11 | 1338 | |||
AK297088.1 | - | 2 | 1165 | |||
AK297336.1 | - | 9 | 1013 | |||
AK301918.1 | - | 6 | 735 | |||
AL355695.1 | - | 1 | 234 | |||
BC000116.1 | - | 9 | 1082 | 3350789 | ||
BC000430.2 | - | 10 | 1181 | 2819828 | ||
BC004359.1 | - | 10 | 1181 | 2819828 | ||
BC103745.1 | - | 10 | 1213 | 6009274 | ||
BX364250.2 | - | 7 | 810 | |||
D79996.1 | - | 10 | 1180 |