Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCAPD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014865.3 | NP_055680.3 | 13 | 1554 | ||
GenBank mRNA | AK022511.1 | - | 13 | 1554 | ||
AK125155.1 | - | 10 | 1209 | |||
AK128354.1 | - | 13 | 1466 | |||
AK303310.1 | - | 12 | 1529 | |||
BC028182.1 | - | 13 | 1566 | 5395775 | ||
D63880.1 | - | 13 | 2307 |