Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MON1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286639.1 | NP_001273568.1 | 3 | 627 | ||
NM_001286640.1 | NP_001273569.1 | 2 | 469 | |||
NM_014940.3 | NP_055755.1 | 4 | 954 | |||
GenBank mRNA | AB020679.1 | - | 4 | 638 | ||
AF442486.1 | - | 3 | 618 | |||
AK023896.1 | - | 4 | 682 | |||
AK128411.1 | - | 1 | 496 | |||
AK293392.1 | - | 2 | 189 | |||
AK296467.1 | - | 3 | 354 | |||
BC024277.1 | - | 4 | 934 | 4843859 |