Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015079.5 | NP_055894.6 | 9 | 2199 | ||
NM_144572.1 | NP_653173.1 | 9 | 2199 | |||
XM_011521387.2 | XP_011519689.1 | 8 | 2144 | |||
XM_017022022.1 | XP_016877511.1 | 11 | 2220 | |||
GenBank mRNA | AB028978.1 | - | 9 | 2062 | ||
AK000173.1 | - | 2 | 1166 | |||
BC033712.1 | - | 4 | 982 | |||
BC140766.1 | - | 8 | 1844 | 9021634 | ||
BC144482.1 | - | 8 | 1844 | 9053009 | ||
BC152465.1 | - | 9 | 2062 | 8862601 |