Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MPRIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015134.3 | NP_055949.2 | 3 | 307 | ||
NM_201274.3 | NP_958431.2 | 3 | 307 | |||
XM_005256563.4 | XP_005256620.1 | 3 | 612 | |||
XM_005256564.4 | XP_005256621.3 | 3 | 612 | |||
XM_011523761.2 | XP_011522063.1 | 3 | 629 | |||
XM_011523762.2 | XP_011522064.1 | 3 | 612 | |||
XM_011523763.2 | XP_011522065.1 | 3 | 612 | |||
XM_011523764.2 | XP_011522066.1 | 3 | 612 | |||
XM_011523765.2 | XP_011522067.1 | 3 | 612 | |||
XM_011523766.2 | XP_011522068.2 | 3 | 612 | |||
XM_011523767.2 | XP_011522069.2 | 3 | 612 | |||
XM_017024393.1 | XP_016879882.1 | 3 | 612 | |||
GenBank mRNA | AB098507.1 | - | 3 | 274 | ||
AB189741.1 | - | 3 | 218 | |||
AL834513.1 | - | 2 | 107 | |||
AY296247.1 | - | 3 | 218 | |||
BC136479.1 | - | 3 | 298 | 9020468 | ||
BE048871.1 | - | 3 | 307 | |||
CF126021.1 | - | 1 | 163 |