Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AP3D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001261826.1 | NP_001248755.1 | 1, 2 | 327 | ||
NM_003938.6 | NP_003929.4 | 1, 2 | 327 | |||
XM_006722932.2 | XP_006722995.1 | 1, 2 | 390 | |||
GenBank mRNA | AB208804.1 | - | 1, 2 | 311 | ||
AF002163.1 | - | 415 | ||||
BC005142.2 | - | 1, 2 | 155 | |||
BC010065.2 | - | 1, 2 | 306 | 4106961 | ||
BQ227203.1 | - | 1, 2 | 327 | |||
U91930.1 | - | 1, 2 | 313 |