Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLUH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015229.3 | NP_056044.3 | 9, 10 | 1233 | ||
XM_011523768.2 | XP_011522070.1 | 8, 9 | 1161 | |||
GenBank mRNA | AB014564.1 | - | 5, 6 | 622 | ||
AK023003.1 | - | 1, 2 | 80 | |||
AK125717.1 | - | 9, 10 | 1253 | |||
BC063786.1 | - | 3, 4 | 312 | |||
CR627449.1 | - | 7, 8 | 968 |